Läs mer
3-Methoxytyrosine hydrochloride, TRC
Produktkod. 30472610
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30472610 100 mg
1 options
Produktkod. 30472610 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM268760100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Methoxytyrosine hydrochloride
CAS 91013-37-5
Molekylformel C10H13NO4 . HCl
Kvantitet 100 mg
Synonym Tyrosine, 3-methoxy-, hydrochloride (7CI), 3-Methoxy-DL-tyrosine monohydrochloride, 3-Methoxy-4-hydroxy-DL-phenylalanine hydrochloride, 3-O-Methyl-DL-DOPA hydrochloride, DL-4-Hydroxy-3-methoxyphenylalanine hydrochloride, 2-Amino-3-(4-hydroxy-3-methoxy-phenyl)propanoic acid hydrochloride (1:1)
InChI formel InChI=1S/C10H13NO4.ClH/c1-15-9-5-6(2-3-8(9)12)4-7(11)10(13)14;/h2-3,5,7,12H,4,11H2,1H3,(H,13,14);1H
LEDER Cl.COc1cc(CC(N)C(=O)O)ccc1O
IUPAC-namn 2-amino-3-(4-hydroxy-3-methoxyphenyl)propanoic acid;hydrochloride
Molekylvikt (g/mol) 247.67
Formel vikt 247.0611