missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
3-Methyl-1,1-diphenylurea, Mikromol™
Beskrivning
3-Methyl-1,1-diphenylurea
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3-Methyl-1,1-diphenylurea |
| CAS | 13114-72-2 |
| Analyt- eller komponentnamn | 3-Methyl-1,1-diphenylurea |
| Frakt skick | Room Temperature |
| Molekylformel | C14 H14 N2 O |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Temozolomide Related Compound B, 3-Methyl-1,1-diphenylurea, Temozolomide Related Compound B |
| InChI formel | InChI=1S/C14H14N2O/c1-15-14(17)16(12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2-11H,1H3,(H,15,17) |
| LEDER | CNC(=O)N(c1ccccc1)c2ccccc2 |
| IUPAC-namn | 3-methyl-1,1-diphenylurea |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?