Läs mer
3-Methyl-1-(2-(1-piperidinyl)phenyl)butylamine Adipate (1:3), TRC
Produktkod. 30470674
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30470674 500 mg
1 options
Produktkod. 30470674 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM326260500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Methyl-1-(2-(1-piperidinyl)phenyl)butylamine Adipate (1:3)
CAS 1262393-01-0
Molekylformel C16H26N2.3 x C6H10O4
Kvantitet 500 mg
Synonym 3-Methyl-1-(2-(1-piperidinyl)phenyl)butylamine Adipic Acid, 3-Methyl-1-(2-(1-piperidinyl)phenyl)butylammonium Adipate, Hexanedioic Acid compd. with alpha-(2-Methylpropyl)-2-(1-piperidinyl)benzenemethanamine
InChI formel InChI=1S/C16H26N2.C6H10O4/c1-13(2)12-15(17)14-8-4-5-9-16(14)18-10-6-3-7-11-18;7-5(8)3-1-2-4-6(9)10/h4-5,8-9,13,15H,3,6-7,10-12,17H2,1-2H3;1-4H2,(H,7,8)(H,9,10)
LEDER CC(C)CC(N)c1ccccc1N2CCCCC2.OC(=O)CCCCC(=O)O
IUPAC-namn hexanedioic acid;3-methyl-1-(2-piperidin-1-ylphenyl)butan-1-amine
Molekylvikt (g/mol) 392.5
Formel vikt 392.2675