Läs mer
3-Methyl-1,3,5-pentanetriol (Technical Grade), TRC
Produktkod. 30483475
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483475 1 g
30479881 100 mg
30479749 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30483475 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3204201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7564-64-9
Molekylformel C6 H14 O3
Molekylvikt (g/mol) 134.17
Synonym 3-Hydroxy-3-methylpentane-1,5-diol, 3-Methyl-1,3,5-pentanetriol, NSC 361412, Petriol
IUPAC-namn 3-methylpentane-1,3,5-triol
LEDER CC(O)(CCO)CCO
Specifikationer
Kemiskt namn eller material 3-Methyl-1,3,5-pentanetriol
Kvantitet 1 g
InChI formel InChI=1S/C6H14O3/c1-6(9,2-4-7)3-5-8/h7-9H,2-5H2,1H3
Formel vikt 134.0943