Läs mer
3-Methyl-1-phenyl-1H-pyrazol-5-yl Beta-D-Glucopyranosiduronic Acid Methyl Ester 2,3,4-Triacetate, TRC
Produktkod. 30469261
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30469261 100 mg
1 options
Produktkod. 30469261 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM354413100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Methyl-1-phenyl-1H-pyrazol-5-yl beta-D-Glucopyranosiduronic Acid Methyl Ester 2,3,4-Triacetate
CAS 663618-33-5
Molekylformel C23H26N2O10
Kvantitet 100 mg
Synonym (2S,3S,4S,5R,6S)-2-(Methoxycarbonyl)-6-((3-methyl-1-phenyl-1H-pyrazol-5-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl Triacetate
InChI formel InChI=1S/C23H26N2O10/c1-12-11-17(25(24-12)16-9-7-6-8-10-16)34-23-21(33-15(4)28)19(32-14(3)27)18(31-13(2)26)20(35-23)22(29)30-5/h6-11,18-21,23H,1-5H3/t18-,19-,20-,21+,23+/m0/s1
LEDER COC(=O)[C@H]1O[C@@H](Oc2cc(C)nn2c3ccccc3)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(5-methyl-2-phenylpyrazol-3-yl)oxyoxane-2-carboxylate
Molekylvikt (g/mol) 490.46
Formel vikt 490.1587