missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%, Thermo Scientific Chemicals

Catalog Number p-166520
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
89-25-8
C10H10N2O
174.2
MFCD00003138
QELUYTUMUWHWMC-UHFFFAOYSA-N
edaravone, 3-methyl-1-phenyl-2-pyrazolin-5-one, norphenazone, radicut, 1-phenyl-3-methyl-5-pyrazolone, developer z, methylphenylpyrazolone, norantipyrine, c.i. developer 1, phenylmethylpyrazolone
4021
CHEBI:31530
5-methyl-2-phenyl-4H-pyrazol-3-one
CC1=NN(C(=O)C1)C2=CC=CC=C2
This item is not returnable. View return policy
89-25-8
C10H10N2O
174.2
MFCD00003138
QELUYTUMUWHWMC-UHFFFAOYSA-N
edaravone, 3-methyl-1-phenyl-2-pyrazolin-5-one, norphenazone, radicut, 1-phenyl-3-methyl-5-pyrazolone, developer z, methylphenylpyrazolone, norantipyrine, c.i. developer 1, phenylmethylpyrazolone
4021
CHEBI:31530
5-methyl-2-phenyl-4H-pyrazol-3-one
CC1=NN(C(=O)C1)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 127.0°C to 131.0°C
Color Beige to Yellow
Boiling Point 287.0°C (265.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 24, 20
Merck Index 15, 6802
Solubility Information Solubility in water: 3g/L water (20°C). Other solubilities: slightly soluble in alcohol and benzene,insoluble in ether,soluble in acids
Formula Weight 174.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-1-phenyl-2-pyrazolin-5-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'