missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Methyl-1-phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H64548.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15491819

  • 12447.17 SEK / 5g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

3-Methyl-1-phenyl-2-thiohydantoin
206.26
MFCD03721167
3-Methyl-1-phenyl-2-thioxo-4-imidazolidinone

Specifications

3-Methyl-1-phenyl-2-thiohydantoin
Solid
206.26
3-Methyl-1-phenyl-2-thioxo-4-imidazolidinone
5g
95%
MFCD03721167
Videos
SDS