Läs mer
[(3-Methyl-1H-1,2,4-triazol-5-yl)thio]acetic acid, TRC
Produktkod. 30473926
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30473926 10 mg
1 options
Produktkod. 30473926 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM24208510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tiazotic Acid
CAS 64679-65-8
Molekylformel C5 H7 N3 O2 S
Kvantitet 10 mg
Synonym [(3-Methyl-1H-1,2,4-triazol-5-yl)thio]acetic acid, Acetic acid, 2-[(3-methyl-1H-1,2,4-triazol-5-yl)thio]-, Acetic acid, [(5-methyl-1H-1,2,4-triazol-3-yl)thio]- (9CI), 2-[(3-Methyl-1H-1,2,4-triazol-5-yl)thio]acetic acid, NSC 78999, Tiazotic acid
InChI formel InChI=1S/C5H7N3O2S/c1-3-6-5(8-7-3)11-2-4(9)10/h2H2,1H3,(H,9,10)(H,6,7,8)
LEDER Cc1nnc(SCC(=O)O)[nH]1
IUPAC-namn 2-[(5-methyl-4H-1,2,4-triazol-3-yl)sulfanyl]acetic acid
Molekylvikt (g/mol) 173.193
Formel vikt 173.026