Läs mer
3-Methyl-2-oxovaleric acid calcium salt, TRC
Produktkod. 30476426
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30476426 500 mg
1 options
Produktkod. 30476426 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM326433500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Methyl-2-oxovaleric acid calcium salt
CAS 66872-75-1
Molekylformel 2 C6 H9 O3 . Ca
Kvantitet 500 mg
Synonym Pentanoic acid, 3-methyl-2-oxo-, calcium salt (2:1), Pentanoic acid, 3-methyl-2-oxo-, calcium salt (9CI), Pentanoic acid, 3-methyl-2-oxo-, calcium salt, (±)-, Calcium 3-methyl-2-oxopentanoate, calcium bis(3-methyl-2-oxopentanoate), α-keto-Isoleucine calcium, 3-Methyl-2-oxopentanoic acid calcium salt, Calcium (±)-3-methyl-2-oxovalerate
InChI formel InChI=1S/2C6H10O3.Ca/c2*1-3-4(2)5(7)6(8)9;/h2*4H,3H2,1-2H3,(H,8,9);/q;;+2/p-2
LEDER [Ca+2].CCC(C)C(=O)C(=O)[O-].CCC(C)C(=O)C(=O)[O-]
IUPAC-namn calcium;3-methyl-2-oxopentanoate
Molekylvikt (g/mol) 298.35
Formel vikt 298.0729