Läs mer
3-Methyl-2-oxovaleric Acid Sodium Salt, TRC
Produktkod. 30133670
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133670 10 g
30012058 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133670 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32642010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3715-31-9
Molekylformel C6 H9 O3 . Na
Molekylvikt (g/mol) 152.12
Synonym Pentanoic acid, 3-methyl-2-oxo-, sodium salt (1:1), Pentanoic acid, 3-methyl-2-oxo-, sodium salt (9CI), Valeric acid, 3-methyl-2-oxo-, sodium salt (8CI), 3-Methyl-2-oxopentanoic acid sodium salt, Sodium DL-3-methyl-2-oxopentanoate, Sodium α-keto-β-methylvalerate, Sodium (±)-3-methyl-2-oxovalerate
IUPAC-namn sodium;3-methyl-2-oxopentanoate
LEDER [Na+].CCC(C)C(=O)C(=O)[O-]
Specifikationer
Kemiskt namn eller material 3-Methyl-2-oxovaleric acid sodium salt
Kvantitet 10 g
InChI formel InChI=1S/C6H10O3.Na/c1-3-4(2)5(7)6(8)9;/h4H,3H2,1-2H3,(H,8,9);/q;+1/p-1
Formel vikt 152.0449