Läs mer
3-Methyl-3,4-dihydro-1H-quinoxalin-2-one, TRC
Produktkod. 30470332
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470332 100 mg
30478281 10 mg
30473395 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30470332 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM303543100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34070-68-3
Molekylformel C9H10N2O
Molekylvikt (g/mol) 162.19
Synonym 3,4-Dihydro-3-methyl-2(1H)-quinoxalinone, 1,2-Dihydro-2-methyl-3-hydroxyquinoxaline, 3,4-Dihydro-3-methyl-1H-quinoxalin-2-one, 3,4-Dihydro-3-methylquinoxalin-2(1H)-one, 3-Methyl-1,2,3,4-tetrahydroquinoxalin-2-one
LEDER CC1Nc2ccccc2NC1=O
Specifikationer
Kemiskt namn eller material 3-Methyl-3,4-dihydro-1H-quinoxalin-2-one
Kvantitet 100 mg
InChI formel InChI=1S/C9H10N2O/c1-6-9(12)11-8-5-3-2-4-7(8)10-6/h2-6,10H,1H3,(H,11,12)
Formel vikt 162.0793