Läs mer
3-Methyl-3-methyloxy-4-benzyloxy-butanoic Acid Methyl Ester, TRC
Produktkod. 30467037
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30467037 250 mg
1 options
Produktkod. 30467037 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM263590250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Methyl-3-methyloxy-4-benzyloxy-butanoic Acid Methyl Ester
CAS 1797024-70-4
Molekylformel C14H20O4
Kvantitet 250 mg
Synonym 3-Methyl-3-methyloxy-4-(phenylmethoxy)-butanoic Acid Methyl Ester
InChI formel InChI=1S/C14H20O4/c1-14(17-3,9-13(15)16-2)11-18-10-12-7-5-4-6-8-12/h4-8H,9-11H2,1-3H3
LEDER O=C(OC)CC(OC)(C)COCC1=CC=CC=C1
IUPAC-namn methyl 4-(benzyloxy)-3-methoxy-3-methylbutanoate
Molekylvikt (g/mol) 252.31
Formel vikt 252.14