Läs mer
3-Methyl-3-phenyl-2-oxiranecarboxylic Acid Methyl Ester, TRC
Produktkod. 30477194
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30477194 10 mg
1 options
Produktkod. 30477194 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33866510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Methyl-3-phenyl-2-oxiranecarboxylic Acid Methyl Ester
CAS 5441-04-3
Molekylformel C11H12O3
Kvantitet 10 mg
Synonym α,β-Epoxy-β-methyl-hydrocinnamic Acid Methyl Ester, 3-Methyl-3-phenyl-oxiranecarboxylic Acid Methyl Ester
LEDER COC(=O)C1OC1(C)c2ccccc2
Molekylvikt (g/mol) 192.21
Formel vikt 192.0786