Läs mer
3-Methyl-4-oxo-3,4-dihydroimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxylic Acid (Temozolomide Acid), Mikromol™
Produktkod. 18011370
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18011370 25 mg
17959589 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18011370 Leverantör Mikromol Leverantörsnummer MM1480.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Methyl-4-oxo-3,4-dihydroimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxylic Acid (Temozolomide Acid)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Temozolomide Acid
CAS 113942-30-6
Analyt- eller komponentnamn Temozolomide Acid
Frakt skick Room Temperature
Molekylformel C6 H5 N5 O3
Certifikat och efterlevnad ISO 17025
Synonym 3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d]-1,2,3,5-tetrazine-8-carboxylic Acid, 3-Methyl-4-oxo-8-imidazolo[5,1-d][1,2,3,5]tetrazinecarboxylic acid, Temozolomide acid, Temozolomide-8-carboxylic acid, Temozolomide Impurity B
InChI formel InChI=1S/C6H5N5O3/c1-10-6(14)11-2-7-3(5(12)13)4(11)8-9-10/h2H,1H3,(H,12,13)
LEDER CN1N=Nc2c(ncn2C1=O)C(=O)O
IUPAC-namn 3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxylic acid
Molekylvikt (g/mol) 195.14
Kvantitet 25 mg
Formel vikt 195.0392 g/mol