missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Methyl-5-isoxazolemethanol, 97%

Brand:  Alfa Aesar™ H63114.03

Additional Details : CAS Number : 14716-89-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499618

  • 5114.93 SEK / 1g


Estimated Shipment date
from Supplier 25-04-2019
Add to basket

Specifications

Chemical Identifiers

14716-89-3
113.116
OFSDWHRZVRCPBR-UHFFFAOYSA-N
10820493
CC1=NOC(=C1)CO
C5H7NO2
MFCD09859210
3-methylisoxazol-5-yl methanol, 3-methyl-1,2-oxazol-5-yl methanol, 3-methyl-5-isoxazolemethanol, 5-isoxazolemethanol,3-methyl, 3-methylisoxazol-5-yl methan-1-ol, 3-methylisoxazole-5-methanol, 5-isoxazolemethanol, 3-methyl, 3-methyl-5-isoxazolyl methanol, 5-hydroxymethyl-3-methylisoxazole, 5-hydroxymethyl-3-methyl-isoxazole
(3-methyl-1,2-oxazol-5-yl)methanol

Specifications

3-Methyl-5-isoxazolemethanol
14716-89-3
MFCD09859210
OFSDWHRZVRCPBR-UHFFFAOYSA-N
(3-methyl-1,2-oxazol-5-yl)methanol
10820493
Viscous Liquid
1g
C5H7NO2
3-methylisoxazol-5-yl methanol, 3-methyl-1,2-oxazol-5-yl methanol, 3-methyl-5-isoxazolemethanol, 5-isoxazolemethanol,3-methyl, 3-methylisoxazol-5-yl methan-1-ol, 3-methylisoxazole-5-methanol, 5-isoxazolemethanol, 3-methyl, 3-methyl-5-isoxazolyl methanol, 5-hydroxymethyl-3-methylisoxazole, 5-hydroxymethyl-3-methyl-isoxazole
CC1=NOC(=C1)CO
113.116
113.11
97%
Videos
SDS