missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Methyl-6-nitro-1H-indazole, 97%

Brand:  Alfa Aesar™ H63005.03

Additional Details : CAS Number : 6494-19-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489428

  • 3172.05 SEK / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Methyl-6-nitro-1H-indazole
6494-19-5
177.16
MFCD07082704

Specifications

3-Methyl-6-nitro-1H-indazole
Solid
177.16
184° to 188°C
6494-19-5
97%
MFCD07082704
1g
Videos
SDS