Läs mer
3-Methyl Adenosine p-Toluenesulfonate Salt, TRC
Produktkod. 30480764
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480764 5 mg
30467280 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30480764 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2761605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72055-63-1
Molekylformel C11H15N5O4 .C7H8SO3
Molekylvikt (g/mol) 455.5
Synonym Adenosine, 3-methyl-, mono(4-methylbenzenesulfonate) (salt) (9CI), 3-Methyladenosine p-toluenesulfonate, 3-Methyl adenosine 4-toluenesulfonate, 3-Methyl adenosine p-toluenesulfonate salt, 3-Methyladenosine tosylate
LEDER CN1C=NC(=N)c2ncn([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)c12.Cc4ccc(cc4)S(=O)(=O)O
Specifikationer
Kemiskt namn eller material 3-Methyl Adenosine p-Toluenesulfonate Salt
Kvantitet 5 mg
InChI formel InChI=1S/C11H17N5O4.C7H8O3S/c1-15-3-14-9(12)6-10(15)16(4-13-6)11-8(19)7(18)5(2-17)20-11;1-6-2-4-7(5-3-6)11(8,9)10/h3-5,7-8,10-11,17-19H,2,12H2,1H3;2-5H,1H3,(H,8,9,10)/t5-,7-,8-,10?,11-;/m1./s1