missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-beta-styrylboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number p-7030529
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1421061-31-5
C15H21BO2
244.141
MFCD12546181
RYYOODZZGKJVFR-MDZDMXLPSA-N
3-methyl-beta-styrylboronic acid pinacol ester, 2-3-methylstyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-3-methylstyryl-1,3,2-dioxaborolane, e-4,4,5,5-tetramethyl-2-3-methylstyryl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-trans-2-m-tolylvinyl 1,3,2 dioxaborolane, 2-e-2-3-methylphenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 4,4,5,5-tetramethyl-2-e-2-3-methylphenyl ethenyl-1,3,2-dioxaborolane
73554487
4,4,5,5-tetramethyl-2-[(E)-2-(3-methylphenyl)ethenyl]-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C=CC2=CC(=CC=C2)C
This item is not returnable. View return policy
1421061-31-5
C15H21BO2
244.141
MFCD12546181
RYYOODZZGKJVFR-MDZDMXLPSA-N
3-methyl-beta-styrylboronic acid pinacol ester, 2-3-methylstyryl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-3-methylstyryl-1,3,2-dioxaborolane, e-4,4,5,5-tetramethyl-2-3-methylstyryl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-trans-2-m-tolylvinyl 1,3,2 dioxaborolane, 2-e-2-3-methylphenyl ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 4,4,5,5-tetramethyl-2-e-2-3-methylphenyl ethenyl-1,3,2-dioxaborolane
73554487
4,4,5,5-tetramethyl-2-[(E)-2-(3-methylphenyl)ethenyl]-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C=CC2=CC(=CC=C2)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 114°C (0.3 mmHg)
Refractive Index 1.5235
Quantity 1 g
Formula Weight 244.14
Percent Purity 97%
Chemical Name or Material 3-Methyl-beta-styrylboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'