missing translation for 'onlineSavingsMsg'
Läs mer

3-Methyl Cytidine Methosulfate, TRC

Produktkod. 30464948
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464948 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30464948 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM294670250MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 3-Methyl Cytidine Methosulfate
CAS 21028-20-6
Molekylformel C10 H16 N3 O5 . C H3 O4 S
Kvantitet 250 mg
Synonym 3-Methylcytidine Mono(Methyl Sulfate) Salt
InChI formel InChI=1S/C10H15N3O5.CH4O4S/c1-12-6(11)2-3-13(10(12)17)9-8(16)7(15)5(4-14)18-9;1-5-6(2,3)4/h2-3,5,7-9,11,14-16H,4H2,1H3;1H3,(H,2,3,4)/t5-,7-,8-,9-;/m1./s1
LEDER COS(=O)(=O)[O-].C[N+]1=C(N)C=CN([C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C1=O
IUPAC-namn 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3-methylpyrimidin-3-ium-2-one;methyl sulfate
Molekylvikt (g/mol) 369.35
Formel vikt 369.0842
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.