Läs mer
3-Methyl Histamine Dihydrochloride, TRC
Produktkod. 30011784
Change view
Kvantitet:
25 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011784 25 mg
30006214 50 mg
30011331 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011784 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31201025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 36475-47-5
Molekylformel C6 H11 N3 . 2 Cl H
Molekylvikt (g/mol) 198.09
IUPAC-namn 2-(3-methylimidazol-4-yl)ethanamine;dihydrochloride
LEDER Cl.Cl.Cn1cncc1CCN
Specifikationer
Kemiskt namn eller material 3-Methylhistamine Dihydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C6H11N3.2ClH/c1-9-5-8-4-6(9)2-3-7;;/h4-5H,2-3,7H2,1H3;2*1H
Formel vikt 197.0487