Läs mer
3-Methyladipic acid, 99%, Thermo Scientific™
Produktkod. 11429337
Change view
Kvantitet:
25 g
100 g
Förpackningsstorlek:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11429337 25 g 25g
11439337 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11429337 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B21387.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3058-01-3
MDL-nummer MFCD00002804
Specifikationer
Kemiskt namn eller material 3-Methyladipic acid
Smältpunkt 97°C to 102°C
Kokpunkt 229°C to 231°C (30 mmHg)
Molekylformel C12H30N4
Kvantitet 25 g
Synonym 1,1,4,7,10,10-hexamethyltriethylenetetramine, hexamethyltriethylenetetramine, 2,5,8,11-tetramethyl-2,5,8,11-tetraazadodecane, hmteta, 1,2-ethanediamine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethyl, n1,n1'-ethane-1,2-diyl bis n1,n2,n2-trimethylethane-1,2-diamine, n,n,n',n,n',n'-hexamethyltriethylenetetramine, triethylenetetramine, 1,1,4,7,10,10-hexamethyl, 1,1,4,7,10,10-hexamethyl-triethylenetetramine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethylethylenediamine
InChI-nyckel DWFKOMDBEKIATP-UHFFFAOYSA-N
LEDER CN(C)CCN(C)CCN(C)CCN(C)C
IUPAC-namn N'-[2-[2-(dimethylamino)ethyl-methylamino]ethyl]-N,N,N'-trimethylethane-1,2-diamine
Molekylvikt (g/mol) 230.40
PubChem CID 76509
Formel vikt 160.17
Procent renhet 99%