Läs mer
[3-(Methylamino)propyl]triphenylphosphonium Bromide Hydrobromide, TRC
Produktkod. 30469888
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30469888 250 mg -
30482181 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30469888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM287025250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4174-83-8
Molekylformel C22H25BrNP . HBr
Molekylvikt (g/mol) 495.2
Synonym Phosphonium, [3-(methylamino)propyl]triphenyl-, bromide, hydrobromide (1:1:1) (ACI), [3-(Methylamino)propyl]triphenylphosphonium bromide, hydrobromide (7CI), Phosphonium, [3-(methylamino)propyl]triphenyl-, bromide, hydrobromide (8CI, 9CI), [3-(Methylamino)propyl]triphenylphosphonium bromide hydrobromide
IUPAC-namn 3-(methylamino)propyl-triphenylphosphanium;bromide;hydrobromide
LEDER Br.[Br-].CNCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
Specifikationer
Kemiskt namn eller material [3-(Methylamino)propyl]triphenylphosphonium Bromide Hydrobromide
Kvantitet 250 mg
InChI formel InChI=1S/C22H25NP.2BrH/c1-23-18-11-19-24(20-12-5-2-6-13-20,21-14-7-3-8-15-21)22-16-9-4-10-17-22;;/h2-10,12-17,23H,11,18-19H2,1H3;2*1H/q+1;;/p-1
Formel vikt 493.017