missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylbenzenecarboximidamide hydrochloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Linear Formula | CH3C6H4C(=NH)NH2·HCl |
Quantity | 1 g |
Formula Weight | 170.64 |
Percent Purity | 97% |
Chemical Name or Material | 3-Methylbenzenecarboximidamide hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'