missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Methylbenzyl isothiocyanate, 99%

Brand:  Alfa Aesar™ H54893.03

Additional Details : CAS Number : 3696-66-0 Weight : 0.00100kg Transport : UN number : 2922 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15451048

  • 1166.62 SEK / 1g


Estimated Shipment date
from Supplier 20-11-2018
Add to basket

Specifications

Chemical Identifiers

3696-66-0
163.238
NUXFAFWRXGFTQJ-UHFFFAOYSA-N
138010
CC1=CC(=CC=C1)CN=C=S
C9H9NS
MFCD00060410
3-methylbenzyl isothiocyanate, 1-isothiocyanatomethyl-3-methylbenzene, 3-methylbenzylisothiocyanate, benzene,1-isothiocyanatomethyl-3-methyl, 1-isothiocyanatomethyl-3-methyl-benzene, 1-isothiocyanatomethyl-3-methyl-benzene, 1-isothiocyanatomethyl-3-methylbenzene #
1-(isothiocyanatomethyl)-3-methylbenzene

Specifications

3-Methylbenzyl isothiocyanate
127°C (2mmHg)
1g
C9H9NS
UN2922
NUXFAFWRXGFTQJ-UHFFFAOYSA-N
1-(isothiocyanatomethyl)-3-methylbenzene
138010
Liquid
1.09g/mL
Brown
3696-66-0
MFCD00060410
3-methylbenzyl isothiocyanate, 1-isothiocyanatomethyl-3-methylbenzene, 3-methylbenzylisothiocyanate, benzene,1-isothiocyanatomethyl-3-methyl, 1-isothiocyanatomethyl-3-methyl-benzene, 1-isothiocyanatomethyl-3-methyl-benzene, 1-isothiocyanatomethyl-3-methylbenzene #
CC1=CC(=CC=C1)CN=C=S
163.238
163.24
99%
Videos
SDS