Läs mer
3-Methylcrotonyl Glycine, TRC
Produktkod. 30137126
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137126 250 mg
30009556 50 mg
30005738 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137126 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM294540250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 33008-07-0
Molekylformel C7 H11 N O3
Molekylvikt (g/mol) 157.17
Synonym Glycine, N-(3-methyl-1-oxo-2-buten-1-yl)- (ACI), Glycine, N-(3-methyl-1-oxo-2-butenyl)- (9CI), Glycine, N-(3-methylcrotonoyl)- (8CI), N-(3-Methyl-1-oxo-2-buten-1-yl)glycine (ACI), 2-(3-Methylbut-2-enamido)acetic acid, 3-Methylcrotonylglycine, N-(3,3-Dimethylacrylyl)glycine, β-Methylcrotonylglycine, N-(3-Methylcrotonyl)glycine
IUPAC-namn 2-amino-5-methyl-3-oxohex-4-enoic acid
LEDER CC(=CC(=O)C(N)C(=O)O)C
Specifikationer
Kemiskt namn eller material 3-Methylcrotonyl Glycine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C7H11NO3/c1-4(2)3-5(9)6(8)7(10)11/h3,6H,8H2,1-2H3,(H,10,11)
Kvantitet 250 mg
Formel vikt 157.0739
Procent renhet >95%