Learn More
3-Methylcyclopentanone, 99%
Product Code. 11391767
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391767 5 g 5g
11381767 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11391767 Supplier Thermo Scientific Alfa Aesar Supplier No. L04940.06
 Restricted Item
I lager
Add to basket
CAS: 1757-42-2 | C6H10O | 98.145 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1757-42-2
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00001416
InChI Key AOKRXIIIYJGNNU-UHFFFAOYSA-N
Synonym 3-methylcyclopentanone, cyclopentanone, 3-methyl, dl-3-methylcyclopentanone, 3-methyl-1-cyclopentanone, cyclopentanone, 3-methyl-, r, 3-methylcyclopentanone, +/-, acmc-20apjx, 3-methyl cyclopentanone, 3-methyl-cyclopentanone, acmc-1ba4k
PubChem CID 15650
IUPAC Name 3-methylcyclopentan-1-one
SMILES CC1CCC(=O)C1
Specifications
Melting Point -58°C
Density 0.914
Boiling Point 144°C to 146°C
Flash Point 36°C (96°F)
Refractive Index 1.434
Quantity 5 g
UN Number UN1224
Beilstein 1305074
Formula Weight 98.15
Percent Purity 99%
Chemical Name or Material 3-Methylcyclopentanone