Läs mer
3-Methylglutaconic Acid, TRC
Produktkod. 30133604
Change view
Kvantitet:
50 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133604 5 g
30010748 50 mg
30005323 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133604 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3115655G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5746-90-7
Molekylformel C6H8O4
Molekylvikt (g/mol) 144.13
Synonym 2-Pentenedioic acid, 3-methyl-, Glutaconic acid, 3-methyl- (6CI,7CI,8CI), 3-Methyl-2-pentenedioic acid, 3-Methylglutaconic acid, NSC 249232, β-Methylglutaconic acid
IUPAC-namn 3-methylpent-2-enedioic acid
LEDER CC(=CC(=O)O)CC(=O)O
Specifikationer
Kemiskt namn eller material 3-Methylglutaconic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C6H8O4/c1-4(2-5(7)8)3-6(9)10/h2H,3H2,1H3,(H,7,8)(H,9,10)
Formel vikt 144.0423
Procent renhet >95%