missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylhexanoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Boiling Point | 215.0°C to 216.0°C |
Flash Point | 102°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96.0 to max. 104% min. (Acid) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 130.19 |
Percent Purity | 97% |
Chemical Name or Material | 3-Methylhexanoic acid |
Safety and Handling
Product Identifier
- 3-Methylhexanoic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'