Läs mer
3-(Methylnitrosamino)propanal, TRC
Produktkod. 30470917
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30470917 10 mg
30465541 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30470917 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32571010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 85502-23-4
Molekylformel C4 H8 N2 O2
Molekylvikt (g/mol) 116.12
Synonym Propanal, 3-(methylnitrosoamino)- (9CI, ACI), 3-(Methylnitrosoamino)propanal (ACI), 3-(Methylnitrosoamino)propionaldehyde, 3-(N-Nitrosomethylamino)propionaldehyde
IUPAC-namn N-methyl-N-(3-oxopropyl)nitrous amide
LEDER CN(CCC=O)N=O
Specifikationer
Kemiskt namn eller material 3-(Methylnitrosamino)propanal
Kvantitet 10 mg
InChI formel InChI=1S/C4H8N2O2/c1-6(5-8)3-2-4-7/h4H,2-3H2,1H3
Formel vikt 116.0586