Läs mer
3-Methyloxindole, TRC
Produktkod. 30475386
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475386 1 g
30469987 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30475386 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3206601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1504-06-9
Molekylformel C9H9NO
Molekylvikt (g/mol) 147.17
Synonym 1,3-Dihydro-3-methyl-2H-indol-2-one, 3-Methyl-2-indolinone
IUPAC-namn 3-methyl-1,3-dihydroindol-2-one
LEDER CC1C(=O)Nc2ccccc12
Specifikationer
Kemiskt namn eller material 3-Methyloxindole
Kvantitet 1 g
InChI formel InChI=1S/C9H9NO/c1-6-7-4-2-3-5-8(7)10-9(6)11/h2-6H,1H3,(H,10,11)
Formel vikt 147.0684