Läs mer
3-Methylpentyl Chloroformate, TRC
Produktkod. 30007987
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007987 5 g
30005108 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007987 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3247255G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1215109-09-3
Molekylformel C7H13ClO2
Molekylvikt (g/mol) 164.63
Synonym Carbonochloridic Acid 3-Methylpentyl Ester
LEDER ClC(OCCC(C)CC)=O
Specifikationer
Kemiskt namn eller material 3-Methylpentyl Chloroformate
Kvantitet 5 g
InChI formel InChI=1S/C7H13ClO2/c1-3-6(2)4-5-10-7(8)9/h6H,3-5H2,1-2H3
Formel vikt 164.0604