Läs mer
3-Methylpyridazine, 99%, Thermo Scientific™
Produktkod. 11458046
Change view
Kvantitet:
5 g
25 g
100 g
Förpackningsstorlek:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11458046 100 g 100g
11438046 5 g 5g
11448046 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11458046 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A14097.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1632-76-4
Molekylformel C5H6N2
Molekylvikt (g/mol) 94.117
MDL-nummer MFCD00006469
InChI-nyckel MXDRPNGTQDRKQM-UHFFFAOYSA-N
Synonym pyridazine, 3-methyl, methyldiazine, pyridazine, methyl, methylpyridazine, 3-methyl pyridazine, pubchem17696, 3-methylpyridazin, acmc-1bp1x, 3-methylpyridazine, ksc181a2j
PubChem CID 74208
IUPAC-namn 3-methylpyridazine
LEDER CC1=NN=CC=C1
Specifikationer
Kemiskt namn eller material 3-Methylpyridazine
Densitet 1.03
Kokpunkt 213°C to 214°C
Flampunkt 87°C (188°F)
Brytningsindex 1.514
Kvantitet 100 g
Beilstein 105691
Formel vikt 94.12
Procent renhet 99%