Läs mer
(3-Methylpyridin-2-yl)methanol, TRC
Produktkod. 30475424
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475424 10 mg
30466073 100 mg
30472409 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30475424 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33793510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 63071-09-0
Molekylformel C7H9NO
Molekylvikt (g/mol) 123.15
Synonym 2-(Hydroxymethyl)-3-methylpyridine, 3-Methylpyridine-2-methanol, 3-Methyl-2-pyridinemethanol
LEDER Cc1cccnc1CO
Specifikationer
Kemiskt namn eller material (3-Methylpyridin-2-yl)methanol
Kvantitet 10 mg
InChI formel InChI=1S/C7H9NO/c1-6-3-2-4-8-7(6)5-9/h2-4,9H,5H2,1H3
Formel vikt 123.0684