Läs mer
3'-N-[[4-(Acetylamino)phenyl]sulfonyl]-3'-N-demethyl Azithromycin, TRC
Produktkod. 17941955
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17941955 100 mg
17904315 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17941955 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA168295100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 612069-30-4
Molekylformel C45 H77 N3 O15 S
Molekylvikt (g/mol) 932.17
Synonym 1-Oxa-6-azacyclopentadecan-15-one, 11-[[3-[[[4-(acetylamino)phenyl]sulfonyl]methylamino]-3,4,6-trideoxy-β-D-xylo-hexopyranosyl]oxy]-13-[(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10-trihydroxy-3,5,6,8,10,12,14-heptamethyl-, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-, 3'-N-[[4-(acetylamino)phenyl]sulfonyl]-3'-N-demethylazithromycin, Azithromycin Related Compound H (USP)
IUPAC-namn N-[4-[[(2S,3R,4S,6R)-2-[[(2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,6,8,10,12,14-heptamethyl-15-oxo-1-oxa-6-azacyclopentadec-11-yl]oxy]-3-hydroxy-6-methyloxan-4-yl]-methylsulfamoyl]phenyl]acetamide
LEDER CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)S(=O)(=O)c4ccc(NC(=O)C)cc4)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O
Specifikationer
Kemiskt namn eller material 3'-N-[[4-(acetylamino)phenyl]sulfonyl]-3'-N-demethylazithromycin
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C45H77N3O15S/c1-15-34-45(11,55)38(51)28(6)47(12)23-24(2)21-43(9,54)40(26(4)37(27(5)41(53)61-34)62-35-22-44(10,58-14)39(52)29(7)60-35)63-42-36(50)33(20-25(3)59-42)48(13)64(56,57)32-18-16-31(17-19-32)46-30(8)49/h16-19,24-29,33-40,42,50-52,54-55H,15,20-23H2,1-14H3,(H,46,49)/t24-,25-,26+,27-,28-,29+,33+,34-,35+,36-,37+,38-,39+,40-,42+,43-,44-,45-/m1/s1
Formel vikt 931.5075
Procent renhet >95%