Läs mer
3-(N-Acetyl-L-cystein-S-yl) Acetaminophen, TRC
Produktkod. 30004570
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004570 10 mg
30004302 2.5 mg
30003792 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004570 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP19110010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52372-86-8
Molekylformel C13H16N2O5S
Molekylvikt (g/mol) 312.34
Synonym N-Acetyl-S-[5-(acetylamino)-2-hydroxyphenyl]-L-Cysteine, PCM-M, 3-[(5-Acetamido-2-Hydroxyphenyl)thio]-N-Acetylalanine, Acetaminophen Mercapturate, N-Acetyl-2-(N-Acetyl-L-Cysteinyl)-4-Aminophenol
LEDER OC1=C(SC[C@H](NC(C)=O)C(O)=O)C=C(NC(C)=O)C=C1
Specifikationer
Kemiskt namn eller material 3-(N-Acetyl-L-cystein-S-yl) Acetaminophen
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C13H16N2O5S/c1-7(16)14-9-3-4-11(18)12(5-9)21-6-10(13(19)20)15-8(2)17/h3-5,10,18H,6H2,1-2H3,(H,14,16)(H,15,17)(H,19,20)/t10-/m0/s1
Kvantitet 10 mg
Formel vikt 312.078
Procent renhet >95%
Frakt skick Dry Pack