Learn More
Alfa Aesar™ 3-n-Butyl-1-methyl-2-thiohydantoin, 95%
Brand:  Alfa Aesar™ H63046.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15449498
  • 2767.11 SEK / 1g
Estimated Shipment date
from Supplier 29-08-2018
Add to basket
Specifications
Chemical Identifiers
3-n-Butyl-1-methyl-2-thiohydantoin
MFCD27665228
3-n-Butyl-1-methyl-2-thioxo-4-imidazolidinone
Specifications
3-n-Butyl-1-methyl-2-thiohydantoin
MFCD27665228
3-n-Butyl-1-methyl-2-thioxo-4-imidazolidinone
Solid
95%
1g
186.27