Learn More
3-N-(Methanesulfonamido)phenylboronic acid, 95%, Thermo Scientific™
Catalog Number p-7077468
Click to view available options
Quantity:
1 g
5 g
148355-75-3
C7H10BNO4S
215.04
XUIQQIRLFMCWLN-UHFFFAOYSA-N
3-methylsulfonylamino phenylboronic acid, 3-methylsulfonamido phenylboronic acid, 3-methanesulfonylaminophenylboronic acid, 3-methylsulfonylaminophenylboronic acid, 3-methanesulfonamidophenyl boronic acid, 3-methanesulfonamido phenyl boronic acid, 3-methanesulfonamidophenylboronic acid, 3-methylsulphonylamino benzeneboronic acid, n-3-methanesulfonamidephenylboronic acid, 3-methylsulfonylamino benzeneboronic acid
2773535
[3-(methanesulfonamido)phenyl]boronic acid
B(C1=CC(=CC=C1)NS(=O)(=O)C)(O)O
148355-75-3
C7H10BNO4S
215.04
XUIQQIRLFMCWLN-UHFFFAOYSA-N
3-methylsulfonylamino phenylboronic acid, 3-methylsulfonamido phenylboronic acid, 3-methanesulfonylaminophenylboronic acid, 3-methylsulfonylaminophenylboronic acid, 3-methanesulfonamidophenyl boronic acid, 3-methanesulfonamido phenyl boronic acid, 3-methanesulfonamidophenylboronic acid, 3-methylsulphonylamino benzeneboronic acid, n-3-methanesulfonamidephenylboronic acid, 3-methylsulfonylamino benzeneboronic acid
2773535
[3-(methanesulfonamido)phenyl]boronic acid
B(C1=CC(=CC=C1)NS(=O)(=O)C)(O)O
Melting Point 255.0°C to 258.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 215.04
Percent Purity 95%
Chemical Name or Material 3-N-(Methanesulfonamido)phenylboronic acid
compliance-icons
Product Identifier
  • 3-N-(Methanesulfonamido)phenylboronic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.