Läs mer
3-(N-Methylpiperidinyl)indole Hydrochloride, TRC
Produktkod. 30480768
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480768 10 mg
30483851 50 mg
30481978 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30480768 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32485210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1956385-76-4
Molekylformel C14 H18 N2 . Cl H
Molekylvikt (g/mol) 250.767
Synonym 3-(1-Methyl-4-piperidinyl)-1H-indole Hydrochloride, 1H-Indole, 3-(1-methyl-4-piperidinyl)-, hydrochloride (1:1), 3-(1-Methyl-4-piperidinyl)-1H-indole hydrochloride, 3-(1-Methylpiperidin-4-yl)-1H-indole hydrochloride
IUPAC-namn 3-(1-methylpiperidin-4-yl)-1H-indole;hydrochloride
LEDER Cl.CN1CCC(CC1)c2c[nH]c3ccccc23
Specifikationer
Kemiskt namn eller material 3-(N-Methylpiperidinyl)indole Hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C14H18N2.ClH/c1-16-8-6-11(7-9-16)13-10-15-14-5-3-2-4-12(13)14;/h2-5,10-11,15H,6-9H2,1H3;1H
Formel vikt 250.124