missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-n-Propyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63060.06

Additional Details : CAS Number : 65710-56-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15499528

  • 14272.98 SEK / 5g


Estimated Shipment date
from Supplier 25-04-2019
Add to basket

Specifications

Chemical Identifiers

65710-56-7
158.219
IHKDRPOYGQYGPI-UHFFFAOYSA-N
4769895
CCCN1C(=O)CNC1=S
C6H10N2OS
MFCD07432487
3-propyl-2-thioxoimidazolidin-4-one, 1-propyl-2-sulfanyl-4,5-dihydro-1h-imidazol-5-one, 2-mercapto-3-propyl-3,5-dihydro-4h-imidazol-4-one, 3-propyl-2-thiohydantoin, 3-propyl-2-thioxo-4-imidazolidinone, 2-thioxo-3-propylimidazolidine-4-one
3-propyl-2-sulfanylideneimidazolidin-4-one

Specifications

3-n-Propyl-2-thiohydantoin
65710-56-7
MFCD07432487
3-propyl-2-thioxoimidazolidin-4-one, 1-propyl-2-sulfanyl-4,5-dihydro-1h-imidazol-5-one, 2-mercapto-3-propyl-3,5-dihydro-4h-imidazol-4-one, 3-propyl-2-thiohydantoin, 3-propyl-2-thioxo-4-imidazolidinone, 2-thioxo-3-propylimidazolidine-4-one
CCCN1C(=O)CNC1=S
158.219
158.22
95%
5g
C6H10N2OS
776219
IHKDRPOYGQYGPI-UHFFFAOYSA-N
3-propyl-2-sulfanylideneimidazolidin-4-one
4769895
Solid
Videos
SDS