Läs mer
3-Nitro-D-tyrosine, TRC
Produktkod. 30477275
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477275 100 mg
30476841 50 mg
30472389 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30477275 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN503883100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32988-39-9
Molekylformel C9H10N2O5
Molekylvikt (g/mol) 226.19
Synonym D-Tyrosine, 3-nitro- (9CI, ACI), 3-Nitro-D-tyrosine (ACI), Tyrosine, 3-nitro-, D- (8CI), (2R)-2-Amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid, (2R)-2-Azaniumyl-3-(4-hydroxy-3-nitrophenyl)propanoate, (R)-2-Amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid
IUPAC-namn (2R)-2-amino-3-(4-hydroxy-3-nitrophenyl)propanoic acid
LEDER N[C@H](Cc1ccc(O)c(c1)[N+](=O)[O-])C(=O)O
Specifikationer
Kemiskt namn eller material 3-Nitro-D-tyrosine
InChI formel InChI=1S/C9H10N2O5/c10-6(9(13)14)3-5-1-2-8(12)7(4-5)11(15)16/h1-2,4,6,12H,3,10H2,(H,13,14)/t6-/m1/s1
Kvantitet 100 mg
Formel vikt 226.059