Learn More
3-Nitrophenyl Ethyl(methyl)carbamate, TRC
Product Code. 30473511
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473511 100 mg -
30464457 10 g 10g
30468003 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30473511 Supplier Toronto Research Chemicals Supplier No. TRCN503470100MG
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1346242-31-6
Molecular Formula C10 H12 N2 O4
Molecular Weight (g/mol) 224.21
Synonym Carbamic acid, N-ethyl-N-methyl-, 3-nitrophenyl ester, 3-Nitrophenyl Ethyl(methyl)carbamate
IUPAC Name (3-nitrophenyl) N-ethyl-N-methylcarbamate
SMILES CCN(C)C(=O)Oc1cccc(c1)[N+](=O)[O-]
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C10H12N2O4/c1-3-11(2)10(13)16-9-6-4-5-8(7-9)12(14)15/h4-7H,3H2,1-2H3
Formula Weight 224.0797
Chemical Name or Material 3-Nitrophenyl Ethyl(methyl)carbamate