missing translation for 'onlineSavingsMsg'
Läs mer

3-Nitropropionic Acid, TRC

Produktkod. 30475001
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30475001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475001 Supplier Toronto Research Chemicals Supplier No. TRCN5197531G

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3-Nitropropionic Acid
CAS 504-88-1
Molekylformel C3H5NO4
Kvantitet 1 g
Synonym BNP, Bovinocidin, Hiptagenic Acid, NSC 64266, β-Nitropropionic Acid
InChI formel InChI=1S/C3H5NO4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)
LEDER O=C(O)CC[N+]([O-])=O
IUPAC-namn 3-nitropropanoic acid
Molekylvikt (g/mol) 119.08
Formel vikt 119.0219
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.