Läs mer
3-O-[(1,1-Dimethylethyl)dimethylsilyl]-1,2:5,6-bis-O-(1-methylethylidene)-Alpha-D-glucofuranose, TRC
Produktkod. 30374677
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30374677 250 mg
1 options
Produktkod. 30374677 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD610870250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-O-[(1,1-Dimethylethyl)dimethylsilyl]-1,2:5,6-bis-O-(1-methylethylidene)-Alpha-D-glucofuranose
CAS 99605-27-3
Molekylformel C18H34O6Si
Kvantitet 250 mg
Synonym Furo[2,3-d]-1,3-dioxole α-D-Glucofuranose Deriv.
InChI formel InChI=1S/C18H34O6Si/c1-16(2,3)25(8,9)24-13-12(11-10-19-17(4,5)21-11)20-15-14(13)22-18(6,7)23-15/h11-15H,10H2,1-9H3/t11-,12-,13+,14-,15-/m1/s1
LEDER CC(C)(C)[Si](C)(C)O[C@H]1[C@H](O[C@@H]2OC(C)(C)O[C@H]12)[C@H]3COC(C)(C)O3
Molekylvikt (g/mol) 374.54