Läs mer
3’-O-(5’-Deoxy-Alpha-D-ribofuranosyl) Capecitabine, TRC
Produktkod. 30136663
Change view
Kvantitet:
0.5 mg
5 mg
Förpackningsstorlek:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136663 5 mg -
30006749 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136663 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2510105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1262133-68-5
Molekylformel C20 H30 F N3 O9
Molekylvikt (g/mol) 475.47
Synonym Cytidine, 5'-deoxy-3'-O-(5-deoxy-alpha-D-ribofuranosyl)-5-fluoro-N-[(pentyloxy)carbonyl]-, 5'-Deoxy-3'-O-(5-deoxy-alpha-D-ribofuranosyl)-5-fluoro-N-[(pentyloxy)carbonyl]cytidine, Pentyl [1-[5-Deoxy-3-O-(5-deoxy-alpha-D-ribofuranosyl)-beta-D-ribofuranosyl]-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl]carbamate
IUPAC-namn pentyl N-[1-[(2R,3R,4S,5R)-4-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]oxy-3-hydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate
LEDER CCCCCOC(=O)NC1=NC(=O)N(C=C1F)[C@@H]2O[C@H](C)[C@@H](O[C@H]3O[C@H](C)[C@@H](O)[C@H]3O)[C@H]2O
Specifikationer
Kemiskt namn eller material 3'-O-(5'-Deoxy-alpha-D-ribofuranosyl) Capecitabine
Kvantitet 5 mg
InChI formel InChI=1S/C20H30FN3O9/c1-4-5-6-7-30-20(29)23-16-11(21)8-24(19(28)22-16)17-14(27)15(10(3)31-17)33-18-13(26)12(25)9(2)32-18/h8-10,12-15,17-18,25-27H,4-7H2,1-3H3,(H,22,23,28,29)/t9-,10-,12-,13-,14-,15-,17-,18-/m1/s1
Formel vikt 475.1966