Läs mer
3-O-Acetylgibberellin A7 Methyl Ester, TRC
Produktkod. 30321619
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30321619 50 mg
1 options
Produktkod. 30321619 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA37756050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-O-Acetylgibberellin A7 Methyl Ester
CAS 5508-48-5
Molekylformel C22H26O6
Kvantitet 50 mg
Synonym 4aalpha,4bbeta-Gibb-3-ene-1alpha,10beta-dicarboxylic Acid, 2beta,4a-Dihydroxy-1-methyl-8-methylene-1,4a-lactone, Methyl Ester, Acetate, Gibberellin A7 Methyl Ester Acetate
InChI formel InChI=1S/C22H26O6/c1-11-9-21-10-13(11)5-6-14(21)22-8-7-15(27-12(2)23)20(3,19(25)28-22)17(22)16(21)18(24)26-4/h7-8,13-17H,1,5-6,9-10H2,2-4H3/t13-,14-,15+,16-,17-,20-,21+,22-/m1/s1
LEDER COC(=O)[C@H]1[C@@H]2[C@@]3(C)[C@@H](OC(=O)C)C=C[C@@]2(OC3=O)[C@@H]4CC[C@@H]5C[C@]14CC5=C
IUPAC-namn methyl (1R,2R,5R,8R,9S,10R,11S,12S)-12-acetyloxy-11-methyl-6-methylidene-16-oxo-15-oxapentacyclo[9.3.2.15,8.01,10.02,8]heptadec-13-ene-9-carboxylate
Molekylvikt (g/mol) 386.44
Formel vikt 386.1729