Läs mer
3-O-Benzyl Estriol, TRC
Produktkod. 30329439
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30329439 250 mg
30338554 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30329439 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB276500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Estriol 3-Benzyl Ether
CAS 18650-87-8
Molekylformel C25 H30 O3
Kvantitet 250 mg
Synonym Estriol 3-Benzyl Ether
InChI formel InChI=1S/C25H30O3/c1-25-12-11-20-19-10-8-18(28-15-16-5-3-2-4-6-16)13-17(19)7-9-21(20)22(25)14-23(26)24(25)27/h2-6,8,10,13,20-24,26-27H,7,9,11-12,14-15H2,1H3/t20-,21-,22+,23-,24+,25+/m1/s1
LEDER C[C@]12CC[C@H]3[C@@H](CCc4cc(OCc5ccccc5)ccc34)[C@@H]1C[C@@H](O)[C@@H]2O
IUPAC-namn (8R,9S,13S,14S,16R,17R)-13-methyl-3-phenylmethoxy-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-16,17-diol
Molekylvikt (g/mol) 378.5
Formel vikt 378.2195