Läs mer
3-O-De(cladinosyl)-8,9,10,11-tetradehydro-9-deoxo-11,12-dideoxy-9,12-epoxy Clarithromycin, TRC
Produktkod. 30008519
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008519 50 mg
30008580 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008519 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22676050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 127157-35-1
Molekylformel C30H51NO8
Molekylvikt (g/mol) 553.73
Synonym 3-O-De(2,6-dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)-8,9,10,11-tetradehydro-9-deoxo-11,12-dideoxy-9,12-epoxy-6-O-methylerythromycin, Clarithromycin EP Impurity K
IUPAC-namn (1S,2R,5R,6S,7S,8R,9R)-8-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-6-hydroxy-9-methoxy-1,5,7,9,11,13-hexamethyl-3,15-dioxabicyclo[10.2.1]pentadeca-11,13-dien-4-one
LEDER O=C1[C@H](C)[C@@H](O)[C@H](C)[C@@H](O[C@H]2[C@H](O)[C@@H](N(C)C)C[C@@H](C)O2)[C@](C)(OC)C/C(C)=C3C(C)=C[C@@](O/3)(C)[C@H](O1)CC
Specifikationer
Kemiskt namn eller material 3-O-De(cladinosyl)-8,9,10,11-tetradehydro-9-deoxo-11,12-dideoxy-9,12-epoxy Clarithromycin
Kvantitet 50 mg
InChI formel InChI=1S/C30H51NO8/c1-12-22-29(7)14-16(2)25(39-29)17(3)15-30(8,35-11)26(19(5)23(32)20(6)27(34)37-22)38-28-24(33)21(31(9)10)13-18(4)36-28/h14,18-24,26,28,32-33H,12-13,15H2,1-11H3/t18-,19+,20-,21+,22-,23+,24-,26-,28+,29+,30-/m1/s1