Learn More
3-O-(E)-Feruloylquinic Acid, TRC
Product Code. 30011217
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011217 5 mg 5mg
30006772 2.5 mg 2.5mg
30004111 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011217 Supplier Toronto Research Chemicals Supplier No. TRCF3089905MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 87099-72-7
Molecular Formula C17H20O9
Molecular Weight (g/mol) 368.34
Synonym trans-3-O-Feruloyl Quinic Acid;
IUPAC Name (1R,3R,4S,5R)-1,3,4-Trihydroxy-5-[[(2E)-3-(4-hydroxy-3-methoxyphenyl)-1-oxo-2-propenyl]oxy]-Cyclohexanecarboxylic Acid
SMILES O[C@@]1(C(O)=O)C[C@@H](OC(/C=C/C2=CC=C(O)C(OC)=C2)=O)[C@@H](O)[C@H](O)C1
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C17H20O9/c1-25-12-6-9(2-4-10(12)18)3-5-14(20)26-13-8-17(24,16(22)23)7-11(19)15(13)21/h2-6,11,13,15,18-19,21,24H,7-8H2,1H3,(H,22,23)/b5-3+/t11-,13-,15+,17-/m1/s1
Chemical Name or Material 3-O-(E)-Feruloylquinic Acid