Läs mer
3-O-Levulinoyl-3,5-dihydroxy Benzoic Acid Acetate, TRC
Produktkod. 30475786
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30475786 100 mg
1 options
Produktkod. 30475786 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL379650100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-O-Levulinoyl-3,5-dihydroxy Benzoic Acid Acetate
CAS 861446-21-1
Molekylformel C14H14O7
Kvantitet 100 mg
Synonym 3-(Acetyloxy)-5-[(1,4-dioxopentyl)oxy]benzoic Acid
InChI formel InChI=1S/C14H14O7/c1-7(15)3-4-11(18)13-10(14(19)20)5-9(17)6-12(13)21-8(2)16/h5-6,17H,3-4H2,1-2H3,(H,19,20)
LEDER O=C(O)C1=CC(O)=CC(OC(C)=O)=C1C(CCC(C)=O)=O
IUPAC-namn 3-acetoxy-5-hydroxy-2-(4-oxopentanoyl)benzoic acid
Molekylvikt (g/mol) 294.26
Formel vikt 294.07