Läs mer
3-O-Methyl-D-glucopyranose, TRC
Produktkod. 30472973
Change view
Kvantitet:
2.5 g
25 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30472973 25 g -
30480146 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30472973 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30945025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3370-81-8
Molekylformel C7H14O6
Molekylvikt (g/mol) 194.18
Synonym 3-O-Methylglucopyranose, 3-O-Methylglucose, NSC 61740, NSC 62383
IUPAC-namn (3R,4S,5R,6R)-6-(hydroxymethyl)-4-methoxyoxane-2,3,5-triol
LEDER CO[C@@H]1[C@@H](O)C(O)O[C@H](CO)[C@H]1O
Specifikationer
Kemiskt namn eller material 3-O-Methyl-D-glucopyranose
Kvantitet 25 g
InChI formel InChI=1S/C7H14O6/c1-12-6-4(9)3(2-8)13-7(11)5(6)10/h3-11H,2H2,1H3/t3-,4-,5-,6+,7?/m1/s1
Formel vikt 194.079